Learn More
BD BBL™ Dehydrated Culture Media: Cystine Tryptic Agar Medium
Catalog No. B11096 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
AOAC
Catalog No. Certifications/Compliance
B11096 AOAC
1 options
This item is not returnable. View return policy
Catalog No. B11096 Supplier BD Supplier No. 211096
Add to Cart
Add to Cart
This item is not returnable. View return policy
Forr maintenance of microorganisms, detection of bacterial motility and with added carbohydrate, for fermentation reactions of fastidious microorganisms
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AOAC
Form Dehydrated
Media Type Cystine Tryptic Agar Medium
Product Type Agar
Formulation Pancreatic digest of Casein 20g/L, L-Cystine 0.5g/L, Sodium Chloride 5g/L, Sodium Sulfite 0.5g/L, Agar 2.5g Phenol red 17mg/L
For Use With (Application) For maintenance of microorganisms, as well as for the detection of bacterial motility and, with added carbohydrate, for fermentation reactions of fastidious microorganisms
Packaging Type Bottle
Quantity 500 g