Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Dermatophyte Test Medium Base

Catalog No. L12330 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L12330 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L12330 Supplier BD Supplier No. 212330
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detection and presumptive identification of dermatophytes from clinical and veterinary specimens

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Form Dehydrated
Media Type Dermatophyte Test Medium Base
Product Type Dehydrated Culture Media
Formulation Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Phenol red 0.2g/L, Cycloheximide 0.5g/L, Agar 20g/L
For Use With (Application) Detection and presumptive identification of dermatophytes from clinical and veterinary specimens
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.