Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Enterococcosel™ Agar

Catalog No. B12205 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CMPH2, ISO, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
B12205 CMPH2, ISO, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B12205 Supplier BD Supplier No. 212205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Bile Esculin Agar with Azide, for the rapid, selective detection and enumeration of enterococci

  • Formulation: Pancreatic digest of Casein 17g/L, Peptic digest of Animal Tissue 3g/L, Yeast extract 5g/L, Oxgall 10g/L, Sodium Chloride 5g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Azide 0.25g/L, Sodium Citrate 1g/L, Agar 13.5g/L

Certifications

CMPH2, ISO, MCM9

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CMPH2, ISO, MCM9
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Enterococcosel Agar
Product Type Agar
Formulation Pancreatic digest of Casein 17g/L, Peptic digest of Animal Tissue 3g/L, Yeast extract 5g/L, Oxgall 10g/L, Sodium Chloride 5g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Azide 0.25g/L, Sodium Citrate 1g/L, Agar 13.5g/L
For Use With (Application) For rapid, selective detection and enumeration of enterococci.1
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.