Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: FTA Hemagglutination Buffer

Catalog No. B11248 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11248 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11248 Supplier BD Supplier No. 211248
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in FTA-ABS test and other serological procedures as a diluent and for washing slide preparations

  • Phosphate Buffered Saline
  • Formulation: Sodium Chloride 7.65g/L, Disodium Phosphate 1268.8mg/L, Monosodium Phosphate 0.1g/L, Monopotassium Phosphate 211.3mg/L
  • pH 7.2
  • Quantity: 500g

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Form Dehydrated
Media Type FTA Hemagglutination Buffer
Product Type Buffer Solution
Formulation Sodium Chloride 7.65g/L, Disodium Phosphate 1268.8mg/L, Monosodium Phosphate 0.1g/L, Monopotassium Phosphate 211.3mg/L
For Use With (Application) Used for the FTA-ABS test and other serological procedures as a diluent and for washing slide preparations
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.