Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: GN Broth

Catalog No. B11279 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11279 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11279 Supplier BD Supplier No. 211279

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for selective enrichment of Salmonella and Shigella

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CCAM, CMPH2, COMPF, MCM9
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type GN Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, D-Mannitol 2g/L, Sodium Citrate 5g/L, Sodium Desoxycholate 0.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L
For Use With (Application) Microbiological examination of food
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.