Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Inhibitory Mold Agar

Catalog No. 1491095 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Inhibitory Mold Agar; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14-910-95 Inhibitory Mold Agar; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-95 Supplier BD Supplier No. 292846
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Culture media for isolation of pathogenic fungi

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BS12, CMPH2, MCM9
Form Dehydrated
Media Type Inhibitory Mold Agar
Product Type Agar
Formulation Pancreatic digest of Casein 3g/L, Peptic digest of Animal Tissue 2g/L, Yeast extract 5g/L, Dextrose 5g/L, Starch 2g/L, Dextrin 1g/L, Chloramphenicol 125mg/L, Sodium Phosphate 2g/L, Magnesium Sulfate 0.8g/L, Ferrous Sulfate 0.04g/L, Sodium Chloride 0.04g/L, Manganese Sulfate 0.16g/L, Agar 15g/L
For Use With (Application) Isolation of pathogenic fungi
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.