Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Kligler Iron Agar

Catalog No. B11317 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11317 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11317 Supplier BD Supplier No. 211317
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For identification of gram-negative enteric bacilli based on fermentation of dextrose and lactose and production of hydrogen sulfide

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, ISO, MCM9
Content And Storage Refrigerate
Form Dehydrated
Media Type Kligler Iron Agar
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Lactose 10g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Thiosulfate 0.5g/L, Agar 15g/L, Phenol red 25mg/L
For Use With (Application) For differentiation of members of the Enterobacteriaceae on the basis of their ability to ferment dextrose and Lactose and to liberate sulfide
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.