Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Liver Veal Agar

Catalog No. DF0059171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0059-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0059-17-1 Supplier BD Supplier No. 259100
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differential medium to detect organisms that decarboxylate or deaminate lysine and produce hydrogen sulfide

Applications:

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Liver Veal Agar
Product Type Agar
Formulation Liver infusion from 50g 9g/L, Veal infusion from 500g 6.4g/L, Proteose Peptone 20g/L, Gelatin 20g/L, Soluble Starch 10g/L, Isoelectric Casein 2g/L, Dextrose 5g/L, Neopeptone 1.3g/L, Tryptone 1.3g/L, Sodium Chloride 5g/L, Sodium Nitrate 2g/L, Agar 15g/L
For Use With (Application) Cultivation of anaerobic bacteria
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.