Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: MacConkey Agar without Crystal Violet

Catalog No. L11393 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L11393 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L11393 Supplier BD Supplier No. 211393
Add to Cart
Add to Cart
This item is not returnable. View return policy

Less selective than MacConkey Agar, to permit growth of staphylococci and enterococci

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey Agar
Product Type MacConkey Agar
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Lactose 10g/L, Bile Salts 5g/L, Sodium Chloride 5g/L, Agar 12g/L, Neutral red 0.05g/L
For Use With (Application) Detection and isolation of gram negative organisms from clinical,1-3 dairy, 4 food, 5-7 Water, 8 pharmaceutical, 9-11 cosmetic, 6, 7 and other industrial source
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.