Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: MacConkey II Agar

Catalog No. B12306 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12306 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B12306 Supplier BD Supplier No. 212306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For isolation of gram-negative enteric bacilli from specimens that may contain swarming strains of Proteus

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

ISO 9001:2008, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, MCM9, SMD, SMWW, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001:2008, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, MCM9, SMD, SMWW, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey II Agar
Product Type Agar
Formulation Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L
For Use With (Application) Isolating of gram negative enteric Bacilli from specimens that may contain swaming strains of proteus
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.