Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: MR-VP Broth

Catalog No. B11383 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Form:
Dehydrated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
B11383 Dehydrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11383 Supplier BD Supplier No. 211383
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For antimicrobial susceptibility testing and isolation of micro-organisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF, ISO, SMD, SMWW, USDA
Form Dehydrated
Media Type MR-VP Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 3.5g/L, Peptic digest of Animal Tissue 3.5g/L, Potassium Phosphate 5g/L, Dextrose 5g/L
For Use With (Application) Antimicrobial susceptibility testing and isolation of microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.