Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Mycophil™ Agar with Low pH

Catalog No. B11450 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11450 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11450 Supplier BD Supplier No. 211450
Add to Cart
Add to Cart

For isolation and enumeration of yeast and molds

TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type Mycophil Agar
Product Type Agar
Formulation Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 18g/L
For Use With (Application) Isolation and enumeration of Yeast and molds
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.