Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Orange Serum Agar

Catalog No. b11486 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11486 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11486 Supplier BD Supplier No. 211486
Only null left
Add to Cart
Edge
Add to Cart

For enumeration and cultivation of microorganisms in citrus juice concentrates and other products

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, COMPF
Content And Storage Refrigerate
Form Dehydrated
Media Type Orange Serum Agar
Product Type Agar
Formulation Orange Serum 10g/L, Yeast extract 3g/L, Pancreatic digest of Casein 10g/L, Dextrose 4g/L, Dipotassium Phosphate 2.5g/L, Agar 15.5g/L
For Use With (Application) Cultivation of aciduric microorganisms, particularly those associated with spoilage of citrus products
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.