Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Phenol Red Lactose Base

Catalog No. B11519 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11519 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11519 Supplier BD Supplier No. 211519
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for determination of fermentation reactions in differentiation of microorganisms

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Lactose Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 10g/L, Sodium Chloride 5g/L, Phenol red 18g/L, Lactose 5g/L
For Use With (Application) Determination of fermentation reactions in the differentiation of microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.