Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: PPLO Broth Base without Crystal Violet

Catalog No. B11458 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11458 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11458 Supplier BD Supplier No. 211458
Add to Cart
Add to Cart

Mycoplasma Broth Base for isolation of micro-organisms

TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type Mycoplasma Broth Base
Product Type Microbiology Media
Formulation Beef heart infusion from (solids) 2g/L, Pancreatic digest of Casein 7g/L, Beef extract 3g/L, Yeast extract 3g/L, Sodium Chloride 5g/L
For Use With (Application) Antimicrobial susceptibility testing and isolation of microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.