Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Schaedler Agar

Catalog No. B12189 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12189 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B12189 Supplier BD Supplier No. 212189
Add to Cart
Add to Cart
This item is not returnable. View return policy

For several media formulations used for recovery of anaerobic microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type Schaedler Agar Base
Product Type Agar
Formulation Pancreatic digest of Casein 8.2g/L, Peptic digest of Animal Tissue 2.5g/L, Papaic digest of Soybean Meal 1g/L, Dextrose 5.8g/L, Yeast extract 5g/L, Sodium Chloride 1.7g/L, Dipotassium Phosphate 0.8g/L, L-Cystine 0.4g/L, Hemin 0.01g/L, Tris (hydroxymethyl) aminomethane 3g/L, Agar 13.5g/L
For Use With (Application) For several media formulations used for the recovery of anaerobic microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.