Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: SIM Medium

Catalog No. B11578 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11578 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11578 Supplier BD Supplier No. 211578
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For differentiating enteric bacilli on the basis of sulfide production, indole formation, and motility

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Content And Storage Refrigerate
Form Dehydrated
Media Type SIM Medium
Product Type Dehydrated Culture Media
Formulation Pancreatic digest of Casein 20g/L, Peptic digest of Animal Tissue 6.1g/L, Ferrous Ammonium Sulfate 0.2g/L, Sodium Thiosulfate 0.2g/L, Agar 3.5g/L
For Use With (Application) To differentiate enteric Bacilli on the basis of Sulfide production, indole formation and motility
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.