Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Sterile Trypticase Soy Broth (Soybean-Casein Digest Broth)

Catalog No. B96264 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B96264 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B96264 Supplier BD Supplier No. 296264
Add to Cart
Add to Cart

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens.

Certifications

AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP
Form Dehydrated
Media Type Trypticase Soy Broth
Product Type Microbiology Media
Sterility Sterile
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.