Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Thioglycollate Medium Brewer Modified

Catalog No. b11716 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11716 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11716 Supplier BD Supplier No. 211716
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Cultivation of obligate anaerobes, microaerophiles and facultative organism

Certifications

CE, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, USP
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Thioglycollate Medium Brewer Modified
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 17.5g/L, Papaic digest of Soybean Meal 2.5g/L, Dextrose 10g/L, Sodium Chloride 5g/L, Sodium Thioglycollate 1g/L, Dipotassium Phosphate 2g/L, Methylene blue 2mg/L, Agar 0.5g/L
For Use With (Application) Cultivation of obligate anaerobes, microaerophiles and facultative organism
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.