Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Trichosel™ Broth, Modified

Catalog No. L11747 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L11747 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L11747 Supplier BD Supplier No. 211747
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detection of microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 25°C
Form Dehydrated
Media Type Trichosel Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 12g/L, Beef extract 2g/L, Yeast extract 5g/L, L-Cysteine HCl 1g/L, Maltose 2g/L, Agar 1g/L, Chloramphenicol 0.1g/L, Methylene blue 3mg/L
For Use With (Application) Isolation of Trichomonas species
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.