Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Trypticase Nitrate Broth

Catalog No. B11299 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11299 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11299 Supplier BD Supplier No. 211299
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Applications:

For identification of microorganisms by means of nitrate reduction and indole test

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Trypticase Nitrate Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 20g/L, Disodium Phosphate 2g/L, Dextrose 1g/L, Agar 1g/L, Potassium Nitrate 1g/L
For Use With (Application) For identification of microorganisms by means of the nitrate reduction and indole test
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.