Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ GN (Gram-Negative) Broth, 8 mL

Catalog No. BB21729 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
No. per Pack:
10
100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
BB21729 10
BB21730 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BB21729 Supplier BD Supplier No. 221729
This item is not returnable. View return policy

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

Used for selective enrichment of Salmonella and Shigella

Inoculate broth as soon as possible after specimens arrive at laboratory.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Pale to Medium, Tan to Yellow, Clear to Slightly Hazy
Broth Type GN (Gram-Negative) Broth
Certifications/Compliance CE, BS12, CCAM, CMPH2, COMPF, MCM9
Color Light Beige
Content And Storage 2°C to 8°C
Additive Sodium Citrate, Sodium Desoxycholate, Dipotassium Phosphate, Monopotassium Phosphate
Detects Salmonella, Shigella
Form Liquid
Manufacturing Origin United States
Media Type GN Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Gram-negative Rods
Test Type Microbiological Tests
Fill Volume 8 mL
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, D-Mannitol 2g/L, Sodium Citrate 5g/L, Sodium Desoxycholate 0.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L
For Use With (Application) Selective enrichment of Salmonella and Shigella
Incubation Conditions 35 ±2°C
Incubation Time 6 to 24 hr.
No. per Pack 10
Packaging Type Tube
Quantity 8 mL
Protein Source Casein, Animal Tissue
Manufacturing Quality CE, BS12, CCAM, CMPH2, COMPF, MCM9
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.