Learn More
Description
- GC Agar supplemented with hemoglobin and BBL IsoVitaleX Enrichment (in place of yeast concentrate) provides an improved medium for the isolation of microorganisms
- Enrichment provides V factor (nicotinamide adenine dinucleotide, NAD) for Haemophilus species
- Also provides vitamins, amino acids, coenzymes, dextrose, ferric ions and other factors which improve the growth of pathogenic Neisseria
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2°C to 8°C |
| Additive | IsoVitaleX™ Enrichment |
| Form | Lyophilized |
| For Use With (Application) | Dehydrated culture media |
| Packaging Type | Vial |
| Quantity | 2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.