Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Lim Broth (Todd Hewitt with CNA) Media

Catalog No. L4392209 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
5 mL
Color:
Beige
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
L4392209 10/Pk. Yellow
L96266 5 mL Beige
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L4392209 Supplier BD Supplier No. 292209
This item is not returnable. View return policy

For selective enrichment of group B Streptococci (Streptococcus agalactiae), especially from genital specimens

  • Available as K tubes

TRUSTED_SUSTAINABILITY

Specifications

Appearance Yellow Liquid
Broth Type Lim Broth (Todd Hewitt with CNA)
Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2, MCM9
Color Yellow
Content And Storage 2°C to 8°C
Detects Tubercle bacilli, Fungi
Form Liquid
Manufacturing Origin United States
Media Type Lim Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Bacterial Pathogens
Test Type Microbial Cultivation
Fill Volume 5 mL
Formulation Heart infusion from (solids) 3.1g/L, Peptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L, Yeast extract 10g/L, Colistin 0.01g/L, Nalidixic Acid 0.015g/L
For Use With (Application) For selective enrichment of group B Streptococci, especially from genital specimens
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Show More Show Less

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.