Learn More
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | CE, BS12, MCM9, SMWW |
| Color | Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Selenite-F Broth |
| Formulation | Pancreatic digest of Casein 5g/L, Lactose 4g/L, Sodium Selenite 4g/L, Sodium Phosphate 10g/L |
| For Use With (Application) | Isolation of Salmonella from feces, urine, Water, foods and other materials of sanitary importance |
| Packaging Type | Tube with Tight Cap |
| Quantity | 100 Pack |
For in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.