Learn More
BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin
Shop All BD Diagnostic Systems ProductsDescription
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, BS12, CLSI, CMPH2, MCM9 |
| Color | Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Thioglycollate Medium |
| Formulation | Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L, Hemin 0.005g/L, Vitamin K1 0.001g/L |
| For Use With (Application) | Isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials |
| Packaging Type | Size A tube with tight cap |
| Quantity | 100 x 10 mL |
For in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.