Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Agar Slants Prepared Media
Shop All BD Diagnostic Systems ProductsDescription
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP |
| Color | Beige |
| Content And Storage | 15°C to 30°C |
| Detects | Fastidious and Non-fastidious Microorganisms, Anaerobic and Aerobic Bacteria, Tubercle bacilli, and Fungi |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Trypticase Soy Agar |
| Product Type | Agar Slants |
| Sterility | Sterile |
| Test Type | Microbial Cultivation |
| Show More |
For in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.