Learn More
BD BBL™ Mycoflask™ Urea Agar Slants, Complete (Christensen) Prepared Media
Shop All BD Diagnostic Systems ProductsDescription
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | CE, BAM, CCAM, ISO, USDA |
| Color | Yellow |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Urea Agar |
| Formulation | Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 0.012g/L, Agar 15g/L |
| For Use With (Application) | For differentiation of organisms, especially Enterobacteriaceae, on basis of Urease production |
| Packaging Type | Glass Tube with Cap |
| Quantity | 10/Pk. |
For in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.