Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Bottled Media: Trypticase Soy Agar (Soybean-Casein Digest Agar)

Catalog No. b99099 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Content And Storage:
2 to 25°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
B99099 2 to 25°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B99099 Supplier BD Supplier No. 299099
Only null left
Add to Cart
Add to Cart

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Isolation and cultivation of non-fastidious and fastidious microorganisms

Certifications

AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Content And Storage 2 to 25°C
Form Liquid
Media Type Trypticase Soy Agar
Product Type Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Isolation and cultivation of non-fastidious and fastidious microorganisms, It is not the medium of choice for anaerobes
Packaging Type Bottle
Quantity 10 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.