Learn More
BD BBL™ Prepared Plated Media: Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™)
Shop All BD Diagnostic Systems ProductsDescription
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Dark Brown |
| Content And Storage | 2°C to 8°C |
| Form | Agar |
| Manufacturing Origin | United States |
| Media Type | Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™) |
| Formulation | Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 12g/L, Hemoglobin 10g/L, IsoVitaleX™ enrichment 10mL |
| For Use With (Application) | Neisseria and Haemophilus species |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.