Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Group A Selective Strep Agar with 5% Sheep Blood (ssA™)

Catalog No. L21779 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Media Type:
Selective Strep Agar, 5% SB
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
L21779 20/Pk. Selective Strep Agar, 5% SB
B21780X 100/Pk. Selective Strep Agar, 5% SB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L21779 Supplier BD Supplier No. 221779
This item is not returnable. View return policy

For the isolation of group A streptococci (S. pyogenes) from throat cultures and other specimens

This media is designed for use in conjunction with Taxo A (bacitracin, 0.04 unit) discs for presumptive identification of S. pyogenes.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Form Solid
Manufacturing Origin United States
Media Type Selective Strep Agar, 5% SB
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, selective Agents 40.2mg/L, Sheep Blood, defibrinated 5%
For Use With (Application) Group A streptococci (S. pyogenes) from throat cultures and other specimens
Packaging Type Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.