Learn More
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Additive | Neutral Red, Crystal Violet, Lactose, Neutral Red Indicator |
| Detects | Coliform Organisms, enteric pathogens |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | MacConkey II Agar |
| Product Type | Prepared Microbiology Plate |
| Target Organism Class | Gram-negative Bacteria |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.