Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Mueller Hinton Agar with 5% Sheep Blood

Catalog No. B21801X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
20/Pk.
24/Pk.
Format:
Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
B21801X 24/Pk. Plate
B21176X 20/Pk. Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B21801X Supplier BD Supplier No. 221801
Add to Cart
Add to Cart
This item is not returnable. View return policy

For antimicrobial disc diffusion susceptibility testing of Streptococcus pneumoniae with selected agents

  • Ready-to-use
  • 100 X 10mm, 100 X 15mm and 150 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Country of Origin United States
Detects Streptococcus pneumoniae
Form Solid
Format Plate
Manufacturing Origin United States
Media Type Mueller Hinton Agar with 5% Sheep Blood
Product Type Prepared Microbiology Plate
Test Type Antimicrobial Susceptibility
Formulation Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L, Defibrinated Sheep Blood 5%
For Use With (Application) Antimicrobial disc diffusion susceptibility testing of Streptococcus pneumoniae with selected agents
Packaging Type Plate
Quantity 24/Pk.
Dimensions (Dia. x H) 150 x 15 mm
Diameter (Metric) 150 mm
Protein Source Beef, Casein, Sheep Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.