Learn More
Description
- Ready-to-use
- Inhibitory to gram-positive organisms
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Orange |
| Content And Storage | 2°C to 8°C |
| Country of Origin | United States |
| Additive | Sodium Desoxycholate, Sodium Thiosulfate, Ferric Ammonium Citrate, Phenol Red |
| Detects | Enteric Pathogens, Shigella species, Salmonella species |
| Form | Liquid |
| Format | Prepared Plate |
| Manufacturing Origin | United States |
| Media Type | XLD Agar |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.