Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Replacement Reagents for Gram Stain Kits
GSA_VA

Catalog No. B12532 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
3.8 L
4 x 250 mL
Product Type:
Basic Fuchsin
Gram Iodine
Gram Iodine (Stabilized)
Safranin
Stain Type:
Basic Fuchsin
Iodine
Safranin
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Stain Type
B12532 3.8 L Safranin Safranin
B12531 4 x 250 mL Safranin Safranin
B12542 4 x 250 mL Gram Iodine Iodine
B12543 3.8 L Gram Iodine (Stabilized) Iodine
B12544 4 x 250 mL Basic Fuchsin Basic Fuchsin
B12545 3.8 L Basic Fuchsin Basic Fuchsin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. B12532 Supplier BD Supplier No. 212532
Add to Cart
Add to Cart

For BBL Gram Stain Kits

  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
  • Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 3.8 L
Certifications/Compliance CE
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Storage Requirements 15° to 30°C
Stain Type Safranin
Product Type Safranin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.