Learn More
Description
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded labels allow quick identification of reagents
- Stabilized Iodine Solution requires no mixing and has a long shelf life
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California
Specifications
Specifications
| Quantity | 4 x 250 mL |
| Certifications/Compliance | CE |
| For Use With (Application) | Used to stain microorganisms from cultures or specimens by the differential gram method |
| Storage Requirements | 15° to 30°C |
| Stain Type | Basic Fuchsin |
| Product Type | Basic Fuchsin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.