Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cephalothin28
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
$284.00
Specifications
Antimicrobial Agents Cephalothin28
Concentration 30 μg
Packaging Type Disc
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
B31271
View Documents
BD
231271
10 Pack
Pack of 10 for $284.00
Only null left
Edge
Add to Cart
B30725
View Documents
BD
230725
1 Pack N/A N/A N/A
Description
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
Cephalothin28
30 μg
Disc
Videos
Safety and Handling
Safety and Handling
ShelfLife : 913 Days
SDS
Documents
Documents
Product Certifications
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.