Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Colistin35
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
$55.50 - $275.00
Specifications
Antimicrobial Agents Colistin35
Concentration 10 μg
Packaging Type Disc
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
B30749
View Documents
BD
230749
1 Pack
Each for $55.50
Only null left
Edge
Add to Cart
B31278
View Documents
BD
231278
10 Pack
Pack of 10 for $275.00
Only null left
Edge
Add to Cart
Description
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
Colistin35
10 μg
Disc
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1460 Days
SDS
Documents
Documents
Product Certifications
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.