Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Piperacillin
Catalog No. B31608 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1/Pk.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31608 1/Pk.
B31609 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B31608 Supplier BD Supplier No. 231608
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Piperacillin
Concentration 100 μg
Packaging Type Disc
Quantity 1/Pk.