Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Streptomycin
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
Specifications
Antimicrobial Agents Streptomycin
Concentration 10μg
Packaging Type Disc
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
B31328
View Documents
BD
231328
10/Pk.
Pack of 10 for $270.00
N/A
Edge
Due to product restrictions, please to purchase or view availability for this product.
B30942
View Documents
BD
230942
1/Pk. Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
View Alternate Product
N/A
Description
Description
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
Specifications
Specifications
Streptomycin
10μg
Disc
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1825 Days
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.