Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Tigecycline

Catalog No. 14959901 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1/Pk.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-959-901 10/Pk.
B232087 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-959-901 Supplier BD Supplier No. 232208
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

  • Clearly marked on both sides with letters and numbers designating agent and concentration

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Tigecycline
Concentration 15 μg
Content And Storage 2°C to 8°C
Packaging Type Disc
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.