Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Thioglycollate Dehydrated Culture Medium without Indicator 135C

Catalog No. B11720 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11720 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11720 Supplier BD Supplier No. 211720
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detection of microorganisms in normally sterile materials, especially those containing mercurial preservatives

Fermentation studies where no oxidation-reduction indicator is required.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CMPH2
Content And Storage -135°C
Form Dehydrated
Media Type Thioglycollate Medium
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, L-Cystine 0.25g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, Sodium Sulfite 0.1g/L
For Use With (Application) For recovery of a wide variety of microorganisms, particularly obligate anaerobes, from clinical specimens and other material
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.