Learn More
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II, I Plate™ Prepared Media
Shop All BD Diagnostic Systems ProductsDescription
Tripticase Soy Agar (TSA II)
- For the visualization of hemolytic reactions produced by many bacterial species
- Selective and differential plating medium for the detection of coliform organisms and enteric pathogens
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light to Medium Red, Medium Rose Tan to Dark Rose Tan Orange Tint |
| Content And Storage | 2°C to 8°C |
| Country of Origin | United States |
| Detects | Escherichia coli, Staphylococcus aureus, Streptococcus pneumoniae, Streptococcus pyogenes, Enterococcus faecalis, Proteus mirabilis, Pseudomonas aeruginosa, Salmonella typhimurium, Shigella dysenteriae |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Trypticase™ Soy Agar (TSA II), MacConkey II Agar |
| Sterility | Sterile |
| Target Organism Class | Fastidious Organisms |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.