Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Calcium Assay Kit

Catalog No. BDB640176 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 Preps
100 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB640176 10 Preps
BDB640178 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB640176 Supplier BD Supplier No. 640176
Only null left
Add to Cart
Add to Cart

Includes

Calcium Indicator, 10X Signal Enhancer

For homogeneous measurement of intracellular Calcium changes

  • Assay involves only one dye addition step and does not require washing
  • Allows easy implementation in high throughput environment

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
For Use With (Application) For homogeneous measurement of intracellular calcium changes
Includes 1 vial calcium indicator,10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X
Quantity 10 Preps
Type Detection Assay Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.