Learn More
Description
Includes
Calcium Indicator, 10X Signal Enhancer
- Assay involves only one dye addition step and does not require washing
- Allows easy implementation in high throughput environment
Specifications
Specifications
| Content And Storage | -20°C |
| Detection Method | Fluorometric |
| For Use With (Application) | For homogeneous measurement of intracellular calcium changes |
| Includes | 10 vials calcium indicator,100mL signal enhancer (10X, lyophilized) |
| Quantity | 100 Preps |
| Type | Detection Assay Kit |
| Druggable Target | Calcium Signaling |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.