Learn More
Description
Includes
Calcium Indicator, 10X Signal Enhancer
- Assay involves only one dye addition step and does not require washing
- Allows easy implementation in high throughput environment
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | For homogeneous measurement of intracellular calcium changes |
| Includes | 1 vial calcium indicator,10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X |
| Quantity | 10 Preps |
| Type | Detection Assay Kit |
Safety and Handling
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.