Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Caspase-3

Catalog No. BDB556472 Shop All BD Biosciences Products
Change view
Click to view available options
Concentration:
0.2mg/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
BDB556472 0.2mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556472 Supplier BD Supplier No. 556472

missing translation for 'validMainframeMsg'

Play key role in normal apoptosis during development induced by the death receptors

Caspases are cysteine proteases that play central role in apoptosis. The caspase family was discovered by searching human cDNA libraries for sequences homologous to ced-3, a C. Each caspase is synthesized as an inactive proenzyme that is processed by cleavage at asparte residues by another protease or by self-proteolysis.

  • Fas and tumor necrosis factor receptor type 1 (TNFR1)
  • One of their substrates is poly (ADP ribose) polymerase (PARP)
  • Cleave PARP from its 116kDa form to 85kDa residual fragment
  • The cleavage site in PARP is C-terminal to Asp-216
  • Utilized as basis for highly specific caspase-3 substrates such as Ac(N-acetyl)-DEVD-AFC (7-amino-4-trifluoromethylcourmarin) and Ac(N-acetyl)-DEVD-AMC (7-amino-4-methylcoumarin) as well as caspase-3 aldehyde inhibitor Ac-DEVD-CHO
  • Storage Buffer: 50mM Tris (pH 8.0) with 100mM NaCl and 50mM imidazole
  • Format: Purified

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2mg/mL
For Use With (Application) Enzyme Assay
Formulation 50mM Tris (pH 8) with 100mM NaCl and 50mM Imidazole
Name Human Caspase-3 Active Form Recombinant Protein
Quantity 5μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Purified

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.