Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Caspase-3, Active Form, mAb Apoptosis Kit : FITC (CPP32)

Catalog No. BDB550480 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550480 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550480 Supplier BD Supplier No. 550480

missing translation for 'validMainframeMsg'

Includes

FITC-conjugated Monoclonal Rabbit Anti-Active Caspase-3 Antibody, BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution, and BD Perm/Wash™ Buffer

For Intracellular staining

  • Format: FITC
  • Clone: C92-605
  • Isotype: Rab IgG
  • Reactivity: Ms, Hu

Intracellular staining, Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Content And Storage 4°C
For Use With (Application) Intracellular Staining
Conjugate FITC
Form Liquid

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.