Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Caspase-3 Active Form, Recombinant Human Protein

Catalog No. BDB556471 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556471 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Avoid multiple freeze-thaws of product
Catalog No. BDB556471 Supplier BD Supplier No. 556471

missing translation for 'validMainframeMsg'

Avoid multiple freeze-thaws of product

Play a key role in cell apoptosis

  • Caspase name indicates two properties: cysteine protease mechanism and ability to cleave after aspartic acid
  • Synthesized as an inactive proenzyme and processed by self-proteolysis to form an active enzyme
  • Processed forms are consist of large (17-22 kDa) and small (10-12 kDa) subunits
  • Used for enzyme assay
  • Format: purified
  • Concentration: 0.2mg/mL
  • Size: 10μg

Education and research purpose

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 μg
Sample Volume 5 μg
Storage Buffer 50mM Tris (pH 8.0) with 100mM NaCl and 50mM Imidazole
Content And Storage -80°C
For Use With (Application) Education, Research
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.