Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Caspase-3 Assay Kit

Catalog No. BDB556485 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556485 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556485 Supplier BD Supplier No. 556485

missing translation for 'validMainframeMsg'

Includes

PBS Buffer, Cell Lysis Buffer, HEPES Buffer, Caspase-3 Fluorogenic Substrate, Caspase-3 Inhibitor

Designed to measure caspase-3 or DEVD-cleaving activity an marker of cells undergoing apoptosis.

  • Used to determine if certain apoptosis pathways involve caspase-3 activation

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Content And Storage 4°C
For Use With (Application) Determine if certain apoptosis pathways involve Caspase-3 activation
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.